Atovaquone 99%

Supplier: Ambeed
Warning

Synonyms: trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione, 2-(trans-4-(4-Chlorophenyl)cyclohexyl)-3-hydroxynaphthalene-1,4-dione

A242112-1MG A242112-250MG A242112-1G A242112-5G A242112-5MG A242112-25G
103791-586EA 7 USD
103791-586 103792-486 103799-524 103811-456 103851-268 76692-304 76692-306 77090-434 77090-436 77090-438 77600-956
Atovaquone 99%
Atovaquone

Precaución: for Research Use Only

Formula: C₂₂H₁₉ClO₃
Boiling Pt: 542.2 °C (760 mm Hg)
Melting Pt: 216…219 °C
Flash Pt: 277.348 °C
Storage Temperature: Freezer
MDL Number: MFCD00889188
CAS Number: 95233-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR