3,4,5-Trimethoxyphenol 97%

Supplier: Ambeed
Warning

Synonyms: Antiarol

A133904-10G A133904-250MG A133904-25G A133904-100G
103801-928EA 23 USD
103801-928 77272-116 77272-118 77314-624 77747-955
3,4,5-Trimethoxyphenol 97%
3,4,5-Trimethoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MW: 184.19
Boiling Pt: 283
Melting Pt: 145…149 °C
Flash Pt: 143.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008389
CAS Number: 642-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR