9-Anthracenecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 9-Anthroic acid, Anthracene-9-carboxylic acid, 9-Anthracenecarboxylic acid, Anthracene-9-carboxylic acid, 9-Carboxyanthracene

A119156-10G A119156-25G
77571-134EA 19.57 USD
77571-134 77571-136
9-Anthracenecarboxylic acid 98%
9-Anthracenecarboxylic acid

Precaución: Sealed in dry, at room temperature.

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 467.5 °C (760 mm Hg)
Melting Pt: 213…217 °C
Flash Pt: 206.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001257
CAS Number: 723-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR