3,4,9,10-Perylenetetracarboxylic dianhydride 95%

Supplier: Ambeed
Warning

Synonyms: Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA

A309952-25G A309952-100G A309952-10G A309952-500G
103835-788EA 5 USD
103835-788 76838-562 77277-492 77277-494 77571-862
3,4,9,10-Perylenetetracarboxylic dianhydride 95%
3,4,9,10-Perylenetetracarboxylic dianhydride

Precaución: for Research Use Only

Formula: C₂₄H₈O₆
Boiling Pt: 755.8±33.0 °C (760 mm Hg)
Melting Pt: 350 °C
Flash Pt: 329.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006916
CAS Number: 128-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR