Amprolium hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Amprolium HCl, 1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride, Amprol, 1-((4-Amino-2-propylpyrimidin-5-yl)methyl)-2-methylpyridin-1-ium chloride hydrochloride, Amprolium hydrochloride, 1-[(4-Amino-2-propyl-5-pyrimidinyl)methyl]-2-methylpyridinium Chloride Hydrochloride, Mepyrium, Thiacoccid

A106091-100G A106091-100MG A106091-25G A106091-500G A106091-5G
103822-748EA 11 USD
103822-748 76687-248 76687-250 76687-252 76687-254 77283-438
Amprolium hydrochloride 97%
Amprolium hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₉ClN₄
MW: 315.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078831
CAS Number: 137-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR