Aminomalonic acid 97%

Supplier: Ambeed
Warning

A176423-1G A176423-5G A176423-100MG A176423-250MG
103807-134EA 160 USD
103807-134 103845-462 77082-816 77082-818 77111-214 77111-216
Aminomalonic acid 97%
Aminomalonic acid

Precaución: for Research Use Only

Formula: C₃H₅NO₄
MW: 119.07 g/mol
Boiling Pt: 402.6 °C (760 mm Hg)
Melting Pt: 240…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00673796
CAS Number: 1068-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR