Aminoguanidine hydrochloride 98%

Supplier: Ambeed
Warning

A287191-100G A287191-100MG A287191-10G A287191-500G A287191-25G
103834-874EA 6 USD
103834-874 76693-162 76693-164 76693-166 76693-168 77305-508
Aminoguanidine hydrochloride 98%
Aminoguanidine hydrochloride

Precaución: for Research Use Only

Formula: CH₇ClN₄
Boiling Pt: 261.4 °C (760 mm Hg)
Melting Pt: 162…166 °C
Flash Pt: 111.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00039074
CAS Number: 1937-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR