AM 580 98%

Supplier: Ambeed
Warning

Synonyms: NSC 608001, CD336, 4-​​[[(5,​​6,​​7,​​8-​​tetrahydro-​​5,​​5,​​8,​​8-​​tetramethyl-​​2-​​naphthalenyl)​carbonyl]​amino]​-​benzoic acid, AM580, 4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido)benzoic acid, RO 40-6055

A184890-1MG A184890-5MG
103791-538EA 24 USD
103791-538 103792-328 103795-066 103799-450 76813-700 77083-996
AM 580 98%
AM 580

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₃
Boiling Pt: 461 °C (760 mm Hg)
Melting Pt: 265…267 °C
Flash Pt: 232.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00673916
CAS Number: 102121-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR