Aluminium trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

Synonyms: Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)

A274248-100G A274248-1G A274248-25G A274248-5G
103813-190EA 9 USD
103813-190 103853-114 76692-906 77092-934 77092-936 77092-938
Aluminium trifluoromethanesulfonate 95%
Aluminium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₃AlF₉O₉S₃
MW: 474.19 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR