2,4,6-Trimethylbenzyl chloride 98%

Supplier: Ambeed
Warning

Synonyms: α-2-Chloroisodurene, α-Chloro-1,2,4,6-tetramethylbenzene, α2-Chloroisodurene, alpha-2-Chloroisodurene, 1-(Chloromethyl)-2,4,6-trimethylbenzene

A213734-25G A213734-5G A213734-100G A213734-10G A213734-500G
103849-032EA 8 USD
103849-032 77087-542 77087-544 77112-518 77112-520 77112-522
2,4,6-Trimethylbenzyl chloride 98%
2,4,6-Trimethylbenzyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃Cl
MW: 168.67 g/mol
Boiling Pt: 244.6 °C (760 mm Hg)
Melting Pt: 36…42 °C
Flash Pt: 106.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000901
CAS Number: 1585-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR