Allylpalladium(II) chloride dimer 97%

Supplier: Ambeed
Warning

Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer

A340926-100MG A340926-250MG A340926-1G A340926-5G A340926-25G
103825-820EA 148 USD
103825-820 103836-798 76693-966 76693-968 76839-818 76839-820 77878-042
Allylpalladium(II) chloride dimer 97%
Allylpalladium(II) chloride dimer

Precaución: for Research Use Only

Formula: C₆H₁₀Cl₂Pd₂
Melting Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044874
CAS Number: 12012-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR