Alloc-Lys(Fmoc)-OH 98%

Supplier: Ambeed
Warning

A259050-1G A259050-5G A259050-100G A259050-250MG A259050-25G
103812-418EA 28 USD
103812-418 103852-296 77091-830 77091-832 77153-386 77153-388 77304-760
Alloc-Lys(Fmoc)-OH 98%
Alloc-Lys(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD09263339
CAS Number: 186350-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR