Aliskiren 98%

Supplier: Ambeed
Warning

A227203-10MG A227203-1MG A227203-25MG A227203-5MG A227203-100MG A227203-250MG
103791-572EA 31 USD
103791-572 103792-448 103792-450 103799-506 103819-306 76691-978 76691-980 76691-982 76691-984 77626-172 77626-174
Aliskiren 98%
Aliskiren

Precaución: for Research Use Only

Formula: C₃₀H₅₃N₃O₆
Boiling Pt: 748.4 °C (760 mm Hg)
Flash Pt: 406.4 °C
Storage Temperature: Freezer
MDL Number: MFCD09839018
CAS Number: 173334-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR