Ricobendazole 97%

Supplier: Ambeed
Warning

Synonyms: Albendazole sulphoxide, N-[6-(propylsulphinyl)-1H-benzimidazol-2-yl]-carbamic acid, methyl ester, Albendazole Oxide, Methyl (6-(propylsulphinyl)-1H-benzo[d]imidazol-2-yl)carbamate

A212431-10G A212431-1MG A212431-25MG A212431-50MG A212431-25G A212431-100G
103848-896EA 31 USD
103848-896 76721-740 76721-742 76721-744 76721-746 77303-246 77600-710
Ricobendazole 97%
Ricobendazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₃S
MW: 281.3 g/mol
Boiling Pt: 357.3 °C (760 mm Hg)
Melting Pt: 154…156 °C
Flash Pt: 169.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797922
CAS Number: 54029-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR