(E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tyrphostin A46, Tyrphostin 46, Tyrphostin AG-99, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide, AG 99, Tyrphostin 46,Tyrphostin A46,Tyrphostin B40, AG-99, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-2-propenamide, 2-Cyano-3-(3,4-dihydroxyphenyl)-2E-propenamide
A1002383-1MG
A1002383-5MG
103791-376EA
19
USD
103791-376
103791-864
103791-866
103799-212
76645-940
77296-606
(E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide 98%
(E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O₃ MW: 204.180 g/mol Boiling Pt: 544.4 °C (760 mm Hg) Flash Pt: 283 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133901 CAS Number: 122520-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...