D-Xylopyranose 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S,5R)-Tetrahydro-2H-pyran-2,3,4,5-tetraol

A103211-100G A103211-25G
103807-856EA 89 USD
103807-856 77670-492 77670-494
D-Xylopyranose 97%
D-Xylopyranose

Precaución: Sealed in dry, at room temperature.

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 415.5 °C (760 mm Hg)
Melting Pt: 148…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00064360
CAS Number: 6763-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR