1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, a-D-Mannose pentaacetate
A116304-100G
A116304-10G
A116304-25G
A116304-5G
A116304-1G
103798-712EA
9
USD
103798-712
103824-572
77189-390
77189-392
77189-394
77297-450
77311-388
1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose 98%
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose
Precaución: for Research Use Only
Formula:
C₁₆H₂₂O₁₁ Boiling Pt: 434.8 °C (760 mm Hg) Melting Pt: 64…75 °C Storage Temperature: Freezer |
MDL Number:
MFCD00069798 CAS Number: 4163-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...