Adamantane-1,3-diyldimethanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A1000923-250MG A1000923-1G
103795-248EA 94 USD
103795-248 103820-964 76645-858 77282-428
Adamantane-1,3-diyldimethanamine dihydrochloride 97%
Adamantane-1,3-diyldimethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₄Cl₂N₂
MW: 267.24 g/mol
Storage Temperature: Ambient
CAS Number: 69887-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR