Adamantane-1,3-diamine dihydrochloride 97%

Supplier: Ambeed
Warning

A208295-250MG A208295-1G A208295-25G A208295-5G
103792-408EA 16 USD
103792-408 103817-868 103848-556 76986-636 77086-864 77086-866 77086-868
Adamantane-1,3-diamine dihydrochloride 97%
Adamantane-1,3-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₀Cl₂N₂
Melting Pt: 308…350 °C
Storage Temperature: Ambient
MDL Number: MFCD00137394
CAS Number: 26562-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR