2-Adamantanone 97%

Supplier: Ambeed
Warning

Synonyms: Adamantan-2-one

A163922-100G A163922-10G A163922-500G A163922-25G A163922-5G
103808-934EA 133 USD
103808-934 103844-056 77247-730 77247-732 77247-734 77300-682 77300-684
2-Adamantanone 97%
2-Adamantanone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Melting Pt: 256…258 °C
Flash Pt: 95.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR