(S)-2-Acetamido-3-methylbutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Ac-Val-OH, Ac-Val-OH, N-Acetyl-L-Valine

A117851-100G A117851-25G A117851-500G
103808-178EA 181 USD
103808-178 77566-154 77566-156 77685-041
(S)-2-Acetamido-3-methylbutanoic acid 98%
(S)-2-Acetamido-3-methylbutanoic acid

Precaución: For research use only. Sealed in dry, store in freezer, under –20 °C.

Formula: C₇H₁₃NO₃
Storage Temperature: Freezer
MDL Number: MFCD00066066
CAS Number: 96-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR