Acridine-9-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Carboxyacridine, 9-Acridinecarboxylic acid
A161207-100MG
A161207-10G
A161207-250MG
A161207-1G
A161207-25G
A161207-5G
A161207-100G
103805-466EA
19
USD
103805-466
103843-704
76783-862
76783-864
76783-866
77300-522
77300-524
77300-526
77871-807
Acridine-9-carboxylic acid 97%
Acridine-9-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₉NO₂ Boiling Pt: 480.4 °C (760 mm Hg) Melting Pt: 290 °C Flash Pt: 244.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009734 CAS Number: 5336-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...