9-Acridone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone
A120093-100G
A120093-10G
A120093-25G
A120093-500G
A120093-1G
A120093-5G
103799-880EA
5
USD
103799-880
77108-522
77108-524
77108-526
77108-528
77297-730
77297-732
9-Acridone 98%
9-Acridone
Precaución: for Research Use Only
Formula:
C₁₃H₉NO Boiling Pt: 355 °C (760 mm Hg) Melting Pt: >300 °C Flash Pt: 155.049 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005019 CAS Number: 578-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...