Ac-Lys(Boc)-OH 96%

Supplier: Ambeed
Warning

Synonyms: Nα-Acetyl-Nepsilon-Boc-L-lysine

A152879-1G A152879-25G
103804-444EA 6 USD
103804-444 103842-616 77299-988 77318-248
Ac-Lys(Boc)-OH 96%
Ac-Lys(Boc)-OH

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₅
MW: 288.344 g/mol
Boiling Pt: 528.8 °C (760 mm Hg)
Melting Pt: 128 °C
Flash Pt: 273.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00236761
CAS Number: 23500-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR