Acetylthiocholine iodide 98%

Supplier: Ambeed
Danger

Synonyms: (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide

A586583-5G A586583-1G
103828-896EA 20 USD
103828-896 103840-040 76844-182 77307-882
Acetylthiocholine iodide 98%
Acetylthiocholine iodide

Precaución: for Research Use Only

Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃I
MW: 289.18 g/mol
Melting Pt: 203…204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011819
CAS Number: 1866-15-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR