Acetimidohydrazide hydrochloride 95%

Supplier: Ambeed
Warning

A376942-100MG A376942-250MG
103826-598EA 354 USD
103826-598 103837-624 76694-400 76694-402
Acetimidohydrazide hydrochloride 95%
Acetimidohydrazide hydrochloride

Precaución: for Research Use Only

Formula: C₂H₈ClN₃
MW: 109.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01746339
CAS Number: 39254-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR