N-Acetyl-D-leucine 99%

Supplier: Ambeed
Warning

Synonyms: N-Acetyle-D-Leucine, N-Ac-D-Leu-OH, Ac-D-Leu-OH, 2-(Acetylamino)-4-methylpentanoic acid

A221188-25G A221188-5G A221188-100G A221188-10G
103849-786EA 10 USD
103849-786 77088-384 77088-386 77685-594 77685-595
N-Acetyl-D-leucine 99%
N-Acetyl-D-leucine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Boiling Pt: 369.7 °C (760 mm Hg)
Melting Pt: 176…177 °C
Flash Pt: 177.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00066069
CAS Number: 19764-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR