9-Vinylanthracene 98%

Supplier: Ambeed
Warning

A189510-250MG A189510-25G A189510-1G A189510-5G
103816-194EA 21 USD
103816-194 103846-786 76690-824 76690-826 77084-556 77084-558
9-Vinylanthracene 98%
9-Vinylanthracene

Precaución: for Research Use Only

Formula: C₁₆H₁₂
Melting Pt: 62…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003576
CAS Number: 2444-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR