9-Phenanthreneboronic acid 97%

Supplier: Ambeed
Warning

A144287-100G A144287-10G A144287-250MG A144287-500G A144287-25G A144287-5G
103803-386EA 6 USD
103803-386 103841-464 76783-092 76783-094 76783-096 76783-098 77299-372 77316-708 77316-710
9-Phenanthreneboronic acid 97%
9-Phenanthreneboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁BO₂
Boiling Pt: 479.5 °C (760 mm Hg)
Melting Pt: 165…170 °C
Flash Pt: 243.796 °C
Storage Temperature: Ambient
MDL Number: MFCD00143524
CAS Number: 68572-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR