Fluorene-9-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 9H-Fluorene-9-carboxylic acid

A439526-10G A439526-250MG A439526-25G A439526-5G A439526-1G
103827-646EA 9 USD
103827-646 103838-712 76725-444 76725-446 76842-468 76842-470 77307-304
Fluorene-9-carboxylic acid 95%
Fluorene-9-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₂
MW: 210.23 g/mol
Boiling Pt: 314.9 °C (760 mm Hg)
Melting Pt: 228…231 °C
Flash Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00001136
CAS Number: 1989-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR