3,9'-Bicarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 9H-3,9'-Bicarbazole

A328047-250MG A328047-25G A328047-1G A328047-5G A328047-500G
103825-490EA 15 USD
103825-490 103836-444 77239-708 77239-710 77306-368 77306-370 77571-934
3,9'-Bicarbazole 97%
3,9'-Bicarbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₂
MW: 332.41 g/mol
Boiling Pt: 598.8±32.0 °C (760 mm Hg)
Flash Pt: 315.9 °C
Storage Temperature: Ambient
MDL Number: MFCD27644690
CAS Number: 18628-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR