9-Fluorenylmethyl carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Carbamic Acid 9-Fluorenylmethyl Ester, Fmoc-NH2, (9H-Fluoren-9-ylmethoxy)carboxamide

A146378-10G A146378-1G A146378-500G A146378-5G A146378-100G
103841-748EA 11 USD
103841-748 77247-128 77247-130 77247-132 77299-516 77549-790
9-Fluorenylmethyl carbamate 98%
9-Fluorenylmethyl carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
Boiling Pt: 459.7 °C (760 mm Hg)
Melting Pt: 201…204 °C
Flash Pt: 242.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00237376
CAS Number: 84418-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR