9-Bromo-1-nonanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Bromononan-1-ol
A308705-25G
A308705-5G
A308705-100G
A308705-10G
A308705-500G
A308705-1G
103814-572EA
5
USD
103814-572
103835-748
76838-510
76838-512
76977-848
76977-850
76977-852
77305-994
9-Bromo-1-nonanol 97%
9-Bromo-1-nonanol
Precaución: For research use only
Formula:
C₉H₁₉BrO Boiling Pt: 277.8 °C (760 mm Hg) Melting Pt: 33…35 °C Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063348 CAS Number: 55362-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...