9-Bromo-9-phenylfluorene 97%
Supplier: Ambeed
|
Danger
|
A207333-250MG
A207333-25G
A207333-5G
A207333-1G
A207333-100G
A207333-10G
103817-786EA
15
USD
103817-786
103848-466
76975-390
77086-740
77086-742
77303-030
77612-040
77671-832
9-Bromo-9-phenylfluorene 97%
9-Bromo-9-phenylfluorene
Precaución: for Research Use Only
Formula:
C₁₉H₁₃Br Boiling Pt: 394.3 °C (760 mm Hg) Melting Pt: 99…101 °C Flash Pt: 172.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075522 CAS Number: 55135-66-5 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...