9-Anthracenemethanol 98%

Supplier: Ambeed
Warning

Synonyms: 9-Anthrylmethanol, Anthracene-9-methanol

A514820-10G A514820-5G
103839-548EA 5 USD
103839-548 76790-046 76843-538
9-Anthracenemethanol 98%
9-Anthracenemethanol

Precaución: for Research Use Only

Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Melting Pt: 162…164 °C
Flash Pt: 196.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001264
CAS Number: 1468-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR