9,9'-Spirobi[9H-fluorene] 98%

Supplier: Ambeed
Warning

A127282-1G A127282-25G A127282-5G A127282-10G A127282-100G A127282-500G
103800-944EA 6 USD
103800-944 77313-188 77313-190 77313-192 77670-798 77881-013 77881-014
9,9'-Spirobi[9H-fluorene] 98%
9,9'-Spirobi[9H-fluorene]

Precaución: for Research Use Only

Formula: C₂₅H₁₆
Boiling Pt: 474.9 °C (760 mm Hg)
Melting Pt: 202 °C
Flash Pt: 236.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08704216
CAS Number: 159-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR