2,7-Dibromo-9,9-di-n-octylfluorene 97%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dioctyl-2,7-dibromofluorene

A307700-25G A307700-5G A307700-100G A307700-10G
103835-694EA 6 USD
103835-694 76838-442 76838-444 77002-614 77002-616
2,7-Dibromo-9,9-di-n-octylfluorene 97%
2,7-Dibromo-9,9-di-n-octylfluorene

Precaución: for Research Use Only

Formula: C₂₉H₄₀Br₂
MW: 548.45 g/mol
Boiling Pt: 567.2 °C (760 mm Hg)
Melting Pt: 59…63 °C
Flash Pt: 341.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03427216
CAS Number: 198964-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR