9,9-Dimethylfluorene 97%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dimethyl-9H-fluorene

A389725-100G A389725-10G A389725-25G A389725-1G A389725-500G
103826-852EA 5 USD
103826-852 103837-894 76694-618 76694-620 76841-264 76841-266 77306-996 77878-123
9,9-Dimethylfluorene 97%
9,9-Dimethylfluorene

Precaución: for Research Use Only

Formula: C₁₅H₁₄
MW: 194.28 g/mol
Boiling Pt: 300.8 °C (760 mm Hg)
Melting Pt: 96 °C
Flash Pt: 138.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00114670
CAS Number: 4569-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR