2-Amino-9,9-dimethylfluorene 98%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dimethyl-9H-fluoren-2-amine

A330603-100G A330603-10G A330603-1G A330603-25G A330603-5G A330603-500G
103836-520EA 9 USD
103836-520 76803-036 76803-038 76803-040 76839-502 76839-504 77668-492
2-Amino-9,9-dimethylfluorene 98%
2-Amino-9,9-dimethylfluorene

Precaución: for Research Use Only

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
Boiling Pt: 374.2 °C (760 mm Hg)
Flash Pt: 194.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09953790
CAS Number: 108714-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR