9,9-Bis(methoxymethyl)fluorene 99%

Supplier: Ambeed
Warning

Synonyms: 9,9-Bis(methoxymethyl)-9H-fluorene

A156007-100MG A156007-250MG A156007-1G A156007-5G A156007-10G
103804-818EA 14 USD
103804-818 103843-020 76984-822 76984-824 77318-762 77318-764 77683-306
9,9-Bis(methoxymethyl)fluorene 99%
9,9-Bis(methoxymethyl)fluorene

Precaución: For research use only.

Formula: C₁₇H₁₈O₂
MW: 254.324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19441380
CAS Number: 182121-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR