9,10-Diphenylanthracene 98%

Supplier: Ambeed
Warning

A244724-100G A244724-10G A244724-1G A244724-25G
103811-598EA 5 USD
103811-598 103851-432 77000-552 77000-554 77090-642 77304-332
9,10-Diphenylanthracene 98%
9,10-Diphenylanthracene

Precaución: for Research Use Only

Formula: C₂₆H₁₈
MW: 330.43 g/mol
Boiling Pt: 469.1 °C (760 mm Hg)
Melting Pt: 245…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00001253
CAS Number: 1499-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR