9,10-Anthraquinone-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A359407-1G
A359407-5G
A359407-100G
103826-226EA
11
USD
103826-226
103837-234
76840-404
76840-406
77668-670
9,10-Anthraquinone-2-carboxylic acid 95%
9,10-Anthraquinone-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₅H₈O₄ Boiling Pt: 518.3 °C (760 mm Hg) Melting Pt: 287…289 °C Flash Pt: 281.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001231 CAS Number: 117-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...