9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Ofloxacin Impurity A

A150088-10G A150088-1G A150088-5G A150088-100G
103804-122EA 17 USD
103804-122 103842-256 77272-960 77299-778 77299-780 77748-042
9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid 97%
9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉F₂NO₄
MW: 281.21 g/mol
Melting Pt: 320…322 °C
Storage Temperature: Ambient
MDL Number: MFCD00226106
CAS Number: 82419-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR