9-(tert-Butoxy)-9-oxononanoic acid 97%

Supplier: Ambeed
Warning

A487079-100MG A487079-250MG A487079-25G A487079-1G A487079-5G
103828-186EA 74 USD
103828-186 103839-260 76649-352 76649-354 76649-356 77307-572 77307-574
9-(tert-Butoxy)-9-oxononanoic acid 97%
9-(tert-Butoxy)-9-oxononanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD30737825
CAS Number: 1290540-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR