9-(4-Bromophenyl)carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 9-(4-Bromophenyl)-9H-carbazole

A203467-10G A203467-500G A203467-5G
103817-466EA 6 USD
103817-466 103848-122 76975-288 76975-290 77086-258
9-(4-Bromophenyl)carbazole 98%
9-(4-Bromophenyl)carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂BrN
MW: 322.21 g/mol
Boiling Pt: 452.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11617969
CAS Number: 57102-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR