9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide 95%

Supplier: Ambeed
Warning

A101676-250MG A101676-1G
103796-086EA 57 USD
103796-086 103821-782 77257-252 77282-796
9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide 95%
9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₁₉BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD27978674
CAS Number: 182438-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR