9-((tert-Butoxycarbonyl)amino)nonanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 9-(Boc-amino)nonanoic acid

A384496-100MG A384496-250MG A384496-25G
103826-766EA 28 USD
103826-766 103837-804 76694-540 76694-542 77626-756
9-((tert-Butoxycarbonyl)amino)nonanoic acid 97%
9-((tert-Butoxycarbonyl)amino)nonanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₇NO₄
MW: 273.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02092968
CAS Number: 173435-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR