8-O-Acetylharpagide 97%

Supplier: Ambeed
Warning

Synonyms: 8-Acetylharpagide, (1S,4aS,5R,7S,7aS)-4a,5-Dihydroxy-7-methyl-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-7-yl acetate, Harpagide 8-acetate, 8-O-Acetylharpagide

A109922-1MG A109922-10MG
103792-008EA 72 USD
103792-008 103792-010 103799-276 77245-648 77310-200
8-O-Acetylharpagide 97%
8-O-Acetylharpagide

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₁₁
MW: 406.38 g/mol
Boiling Pt: 607.2 °C (760 mm Hg)
Flash Pt: 215.9 °C
Storage Temperature: Freezer
MDL Number: MFCD03700750
CAS Number: 6926-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR