8-Methyl-7H-purin-6-amine 98%

Supplier: Ambeed
Warning

A161222-100MG A161222-250MG
103805-468EA 460 USD
103805-468 76985-056 76985-058
8-Methyl-7H-purin-6-amine 98%
8-Methyl-7H-purin-6-amine

Precaución: for Research Use Only

Formula: C₆H₇N₅
MW: 149.15 g/mol
Boiling Pt: 495 °C (760 mm Hg)
Flash Pt: 285.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19237006
CAS Number: 22387-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR