8-Methyl-3,4-dihydroisoquinolin-1(2H)-one 98%

Supplier: Ambeed
Warning

A196193-100MG A196193-250MG A196193-5G A196193-1G
103816-826EA 83 USD
103816-826 76986-292 76986-294 76986-296 77085-396
8-Methyl-3,4-dihydroisoquinolin-1(2H)-one 98%
8-Methyl-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 404.3±45.0 °C (760 mm Hg)
Flash Pt: 237.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11007915
CAS Number: 1082041-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR